Cysteaminiumchlorid, Pierce™
Lieferant: Thermo Fisher Scientific
Synonyme:
2-MEA
20408
PIER20408EA
301
EUR
PIER20408
Cysteaminiumchlorid, Pierce™
Cysteaminiumchlorid
Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₂H₇NS·HCl Molecular Weight: 113,61 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00012904 CAS-Nummer: 156-57-0 EINECS: 205-858-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...