Protein A/G agarose, Pierce™
Lieferant: Thermo Fisher Scientific
20421 20422
PIER20421EA 556 EUR
PIER20421 PIER20422
Protein A/G agarose, Pierce™
Protein-Aufbereitungs Protein Purification Resins
Pierce™ Protein A/G agarose is a affinity resin for antibody purification.
  • Immobilised Protein A/G is ideal for polyclonal IgG purification from nearly any mammalian species
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A/G is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilised Protein A/G
Binding capacity: 9 mg mouse IgG or ≥7 mg human IgG/ml of settled resin