Natriumperiodat, Pierce™
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Thermo Scientific Pierce Sodium meta-Periodate is a gentle oxidizing agent that cleaves cis-diols in carbohydrate sugars to create amine-reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.
- Related applications: Protein Labeling & Crosslinking
Formel:
NaIO₄ Siedepunkt: 300 °C (1013 hPa) Schmelzpunkt: 175 °C Dichte: 3,87 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003534 CAS-Nummer: 7790-28-5 EINECS: 232-197-6 UN: 1479 ADR: 5.1,II Merck Index: 13,08714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...