Sulpho-KMUS, Pierce™
Lieferant: Thermo Fisher Scientific
Thermo Scientific Pierce Sulfo-KMUS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a long spacer arm (16.3 angstroms).
- Reactive groups: sulfo-NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups
- Long aliphatic spacer arm compared to Sulfo-GMBS
- Sulfo-NHS ester reacts with primary amines at pH 7-9, forming a stable amide bond
- Maleimide reacts with -SH groups at pH 6.5-7.5, forming a stable thioether bond
- Non-cleavable
- Water-soluble
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₉H₂₅N₂NaO₉S Lagertemperatur: Kühlschrank |
MDL:
MFCD01863412 CAS-Nummer: 211236-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...