Sulpho-NHS-LC-Biotin, EZ-Link™, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Sulphosuccinimidyl-6-(biotinamido)hexanoate, Biotinamidohexanoic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt

21335
PIER21335EA 426 EUR
PIER21335
Sulpho-NHS-LC-Biotin, EZ-Link™, Pierce™
Sulpho-NHS-LC-Biotin

Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.


  • Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
  • Medium length—spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass


Sulfo-NHS-LC-Biotin is one of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-LC-Biotin is offered in several package sizes and as complete protein labeling kits.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures. Store desiccated at -20°C.

Formel: C₂₀H₂₉N₄NaO₉S₂
Molecular Weight: 556,59 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00078534
CAS-Nummer: 127062-22-0

Bestellen

Spezifikation Testergebnisse

Description EZ-Link Sulfo-NHS-LC-Biotin
Product Type Sulfo-NHS-LC-Biotin
Quantity 100 mg
Product Line EZ-Link™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Label Type Biotin & Analogs
Spacer Mid-Length

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR