Affinity purification beads and magnets, MagnaBind™
Lieferant: Thermo Fisher Scientific
21354 21356 21344 21352 21353 21349 21348
PIER21354EA 506 EUR
PIER21354 PIER21356 PIER21344 PIER21352 PIER21353 PIER21349 PIER21348
Affinity purification beads and magnets, MagnaBind™
Kugeln Magnetkugeln
Magnetic beads are a convenient affinity support for a variety of assays, which allow easy purification of the target without columns or centrifugation. After a binding step in an affinity purification procedure, the magnetic particles are easily and rapidly collected by placing the microcentrifuge tube or reaction vessel next an appropriate rare-earth magnet. MagnaBind™ beads respond rapidly to MagnaBind™ magnets but can be easily dispersed and regathered multiple times (ie. they will not irreversibly aggregate) because they do not have any magnetic memory.
  • Available pre-coated with
More Product Information