EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Bis(2,5-dioxopyrrolidin-1-yl) O,O'-ethane-1,2-diyl disuccinate, EGS, Ethylene glycol disuccinate di(N-succinimidyl) ester, Ethylene glycol bis(succinimidyl succinate), Di(N-succinimidyl) ethylene glycol disuccinate

21565
PIER21565EA 258 EUR
PIER21565
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], Pierce™
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]

Thermo Scientific Pierce DTSSP is a popular water-soluble crosslinker that contains amine-reactive NHS-ester ends around an 8-atom spacer arm, whose central disulfide bond can be cleaved with reducing agents.


  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-EGS
  • Membrane-permeable, allowing for intracellular crosslinking
  • Crosslinks formed are reversible at pH 8.5 using hydroxylamine for 3 to 6 hours at 37°C
  • Lactose dehydrogenase retained 60% of its activity after reversible crosslinking with EGS


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₈H₂₀N₂O₁₂
Molecular Weight: 456,36 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00049057
CAS-Nummer: 70539-42-3

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Cleavable, Long
Form Powder
Quantity 1 g
Type Crosslinker
Solubility DMF, DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR