Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Pierce™

Lieferant: Thermo Fisher Scientific

21566
PIER21566EA 224 EUR
PIER21566
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Pierce™
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate))

Thermo Scientific Pierce Sulfo-EGS is a water-soluble crosslinker that contains amine-reactive sulfo-NHS-ester ends around a 12-atom spacer arm, which can be cleaved by treatment with hydroxylamine at pH 8.5.


  • Reactive groups: sulfo-NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Water-soluble; compare to EGS
  • Membrane-impermeable, allowing for cell surface labeling
  • Crosslinks formed are reversible at pH 8.5 using hydroxylamine for 3 to 6 hours at 37°C


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₈H₁₈O₁₈N₂Na₂S₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00054988
CAS-Nummer: 67410-92-6

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Sulfo-NHS Ester
Spacer Cleavable, Long
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water
Spacer Arm 16.1 Å


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR