Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate) ≥90%, Pierce™
Lieferant: Thermo Fisher Scientific
Synonyme:
Sulfo LC SPDP
Thermo Scientific Pierce Sulfo-LC-SPDP is a water-soluble, long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteines.
- Reactive groups: NHS ester and pyridyldithiol
- Reactive towards: amino and sulfhydryl groups
- Releases a detectable by-product when reacted with a free sulfhydryl group; by measuring the release of pyridine-2-thione at 343 nm, the reaction can be easily followed
- Water soluble, sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
- Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
- Spacer arm is also easily cleaved using reducing SDS-PAGE sample loading buffer
- Cleavable crosslinker allows separation of crosslinked products
- Sulfo-LC-SPDP crosslinker is not membrane permeable, allowing cell surface labeling
- Compare to other varieties of SPDP-type reagents, including pegylated forms
Sulfo-LC-SPDP is Sulfosuccinimidyl 6-(3'-[2-pyridyldithio]-propionamido)hexanoate, a heterobifunctional, thiol-cleavable and membrane impermeable crosslinker. It contains an amine-reactive N-hydroxysuccinimide (NHS) ester that will react with lysine residues to form a stable amide bond. The other end of the spacer arm is terminated in the pyridyl disulfide group that will react with sulfhydryls to form a reversible disulfide bond.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₈H₂₂N₃NaO₈S₃ Molecular Weight: 527,567 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00083162 CAS-Nummer: 169751-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...