LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: LC SPDP

21651
PIER21651EA 576 EUR
PIER21651
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate), Pierce™
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate)

Thermo Scientific Pierce LC-SPDP is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.


  • Reactive groups: NHS ester and pyridyldithiol
  • Reactive towards: amino and sulfhydryl groups
  • Releases a detectable by-product when reacted with a free sulfhydryl group; by measuring the release of pyridine-2-thione at 343 nm, the reaction can be easily followed
  • Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
  • Spacer arm is also easily cleaved using reducing SDS-PAGE sample loading buffer
  • Cleavable crosslinker allows separation of crosslinked products
  • Water-insoluble (dissolve first in DMF or DMSO)
  • SPDP crosslinker is membrane permeable, so crosslinking can be done inside the cells
  • Compare to other varieties of SPDP-type reagents, including pegylated forms


We offer three analogs of SPDP: the standard version (SPDP), a derivative with a longer spacer arm (LC-SPDP), and a sulfonated water-soluble variety (Sulfo-LC-SPDP). SPDP (N-Succinimidyl 3-(2-pyridyldithio)-propionate) and LC-SPDP (Succinimidyl 6-(3-[2-pyridyldithio]-propionamido)hexanoate) are heterobifunctional, thiol-cleavable and membrane permeable crosslinkers. They each contain an amine-reactive N-hydroxysuccinimide (NHS) ester that will react with lysine residues to form a stable amide bond. The other end of the spacer arm is terminated in the pyridyl disulfide group that will react with sulfhydryls to form a reversible disulfide bond.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₈H₂₃N₃O₅S₂
Molecular Weight: 425,522 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00083163
CAS-Nummer: 158913-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR