DMP (Dimethyl pimelimidate dihydrochloride), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: DMP

21666 21667
PIER21666EA 64.5 EUR
PIER21666 PIER21667
DMP (Dimethyl pimelimidate dihydrochloride), Pierce™
DMP (Dimethyl pimelimidate dihydrochloride)

Thermo Scientific Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.


  • Reactive groups: imidoester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Mid-length reagent of three similar imidoester crosslinkers, which include DMA, DMP and DMS
  • Water-soluble
  • Imidoesters react with amines at alkaline pH values (pH 8-10) to form amidine bond
  • Amidine bond retains net charge character of protein primary amine to which it reacts
  • Reversible at high pH values
  • Tool for study of quarternary structure of proteins


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₉H₁₈N₂O₂
Molecular Weight: 259.17 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00038942
CAS-Nummer: 58537-94-3

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Short
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water
Spacer Arm 9.2 Å


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR