AMAS (N-Succinimidyl maleimidoacetate), Pierce™
Lieferant: Thermo Fisher Scientific
Synonyme:
AMAS
22295
PIER22295EA
177
EUR
PIER22295
AMAS (N-Succinimidyl maleimidoacetate), Pierce™
AMAS (N-Succinimidyl maleimidoacetate)
Thermo Scientific Pierce AMAS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a very short spacer arm (4.4 angstroms).
- Reactive groups: NHS ester and maleimide
- Reactive towards: amino and sulfhydryl groups
- Shortest of aliphatic spacer series which includes AMAS, BMPS, GMBS and EMCS
- NHS ester end couples with primary amines at pH 7-9, forming stable amide bonds
- Maleimide reacts with -SH groups at pH 6.5-7.5, forming stable thioether linkages
- Non-cleavable
- Water-insoluble (dissolve first in DMF or DMSO)
- Very short aliphatic spacer has low potential for eliciting an immune response
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₀H₈N₂O₆ Molecular Weight: 252,18 g/mol |
MDL:
MFCD00133523 CAS-Nummer: 55750-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...