BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: BMPH

22297
PIER22297EA 180 EUR
PIER22297
BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt), Pierce™
BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt)

Thermo Scientific Pierce BMPH is a short, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).


  • Reactive groups: maleimide and hydrazide
  • Reactive towards: sulfhydryl groups and carbonyl (aldehyde) groups
  • Short (8.1A), sulfhydryl-to-aldehyde crosslinker with simple spacer arm (noncleavable)
  • Maleimide group reacts with sulfhydryl groups to form stable thioether linkages
  • Hydrazide group conjugates to oxidized sugars of glycoproteins and carbohydrates
  • Use sodium meta-periodate to oxidize glycosylation (e.g., sialic acid) to reactive aldehyde groups
  • Use with EDC to conjugate primary amine of hydrazide group to carboxyl groups


Related Products: EMCH, MPBH, KMUH


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₇H₉N₃O₃·CF₃CO₂H
Molecular Weight: 297,19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863410
CAS-Nummer: 359436-61-6

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Hydrazide, Maleimide
Spacer Short
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR