BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS

22298
PIER22298EA 175 EUR
PIER22298
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester), Pierce™
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)

Thermo Scientific Pierce BMPS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short spacer arm (5.9 Å).


  • Reactive groups: NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • C3-homolog of aliphatic spacer series which includes AMAS, BMPS, GMBS and EMCS
  • NHS ester end couples with primary amines at pH 7-9, forming stable amide bonds
  • Maleimide reacts with -SH groups at pH 6.5-7.5, forming stable thioether linkages
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO)
  • Short aliphatic spacer has low potential for eliciting an immune response


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₁H₁₀N₂O₆
Molecular Weight: 266,21 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043141
CAS-Nummer: 55750-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR