Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester), Pierce™

Lieferant: Thermo Fisher Scientific

22307
PIER22307EA 419 EUR
PIER22307
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester), Pierce™
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)

Thermo Scientific Pierce Sulfo-EMCS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length spacer arm (9.4 angstroms).


  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Sulfo-NHS ester end couples with primary amines at pH 7-9 to form stable amide bonds
  • Maleimide reacts with -SH groups at pH 6.5-7.5, forming stable thioether linkages
  • Non-cleavable
  • Water-soluble (compare to EMCS)
  • Aliphatic spacer offers low potential for eliciting an immune response


Sulfosuccinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate (Sulfo-SMCC) is a non-cleavable and membrane impermeable crosslinker. It contains an amine-reactive N-hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC and SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to ∼10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR