MBS (N-Succinimidyl 3-maleimidobenzoate), Pierce™
Lieferant: Thermo Fisher Scientific
Synonyme:
MBS
22311
PIER22311EA
155
EUR
PIER22311
MBS (N-Succinimidyl 3-maleimidobenzoate), Pierce™
MBS (N-Succinimidyl 3-maleimidobenzoate)
Thermo Scientific Pierce MBS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 angstroms).
- Reactive groups: NHS ester and maleimide
- Reactive towards: amino and sulfhydryl groups
- Non-cleavable
- Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-MBS
- Popular for forming enzyme immunoconjugates
Formel:
C₁₅H₁₀N₂O₆ Molecular Weight: 314,25 g/mol |
MDL:
MFCD00005514 CAS-Nummer: 58626-38-3 |
Spezifikation Testergebnisse
Product Line | Pierce™ |
Labeling Method | Chemical Labeling |
Crosslinker Type | Heterobifunctional |
Reactive Moiety | Maleimide, NHS Ester |
Spacer | Short |
Form | Powder |
Quantity | 50 mg |
Type | Crosslinker |
Solubility | DMF, DMSO |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...