Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Sulfo MBS

22312
PIER22312EA 217 EUR
PIER22312
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt), Pierce™
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Thermo Scientific Pierce Sulfo-MBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 angstroms).


  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Non-cleavable
  • Water-soluble (compare to MBS)
  • Membrane impermeable, allowing cell surface labeling


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₅H₉N₂O₉S·Na
Molecular Weight: 416,295 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01318594
CAS-Nummer: 92921-25-0

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR