Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Sulfo GMBS

22324
PIER22324EA 320 EUR
PIER22324
Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester), Pierce™
Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester)

Thermo Scientific Pierce Sulfo-GMBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short spacer arm (7.3 angstroms).


  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Non-cleavable
  • Water-soluble (compare to GMBS)
  • Has a low potential for illiciting crosslinker immune response, ensuring that the primary response to the antigen-carrier protein conjugate is not diluted by a response against a determinant on the crosslinker
  • GMBS and Sulfo-GMBS are reported to be less immunogenic than SMCC


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₂H₁₂N₂O₉S
Molecular Weight: 360,297 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00153605
CAS-Nummer: 158018-86-1

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR