Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Sulfo SIAB

22327
PIER22327EA 279 EUR
PIER22327
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate), Pierce™
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)

Thermo Scientific Pierce Sulfo-SIAB a water-soluble, mid-length (10.6 angstrom) crosslinker for amine-to-sulfhydryl conjugation via sulfo-N-hydroxysuccinimide (sulfo-NHS) ester and iodoacetyl reactive groups.


  • Reactive groups: sulfo-NHS ester and iodoacetyl
  • Reactive toward: amino and sulfhydryl groups
  • Non-cleavable
  • Water-soluble (compare to SIAB)


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Haloacetyl, Sulfo-NHS Ester
Spacer Mid-Length
Form Powder
Quantity 50 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR