1,1'-(1,6-Hexandiyl)bis(1H-pyrrol-2,5-dion), Pierce™

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: BMH

22330
PIER22330EA 173 EUR
PIER22330
1,1'-(1,6-Hexandiyl)bis(1H-pyrrol-2,5-dion), Pierce™
BMH (1,6-Bis(maleimido)hexane)

Thermo Scientific Pierce BMH is a long-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).


  • Reactive groups: maleimide (both ends)
  • Reactive towards: sulfhydryl groups
  • Long (13.0Å), sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 11-atom spacer arm
  • Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
  • Noncleavable


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₄H₁₆N₂O₄
Molecular Weight: 276,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00047122
CAS-Nummer: 4856-87-5

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Long
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO
Spacer Arm 16.1 Å

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR