BMB (1,4-Bis(maleimido)butane), Pierce™
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
BMB, 1,4-Di-(maleinimido)-butan
22331
PIER22331EA
166
EUR
PIER22331
BMB (1,4-Bis(maleimido)butane), Pierce™
BMB (1,4-Bis-(maleinimido)-butan)
Thermo Scientific Pierce BMB is a mid-length, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).
- Reactive groups: maleimide (both ends)
- Reactive towards: sulfhydryl groups
- Mid-length (10.9Å), sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 9-atom spacer arm
- Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
- Noncleavable
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₂H₁₂N₂O₄ Molecular Weight: 248,24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01318342 CAS-Nummer: 28537-70-4 |
Spezifikation Testergebnisse
Product Line | Pierce™ |
Labeling Method | Chemical Labeling |
Crosslinker Type | Homobifunctional |
Reactive Moiety | Maleimide |
Spacer | Mid-Length |
Form | Solid |
Quantity | 50 mg |
Type | Crosslinker |
Solubility | DMF, DMSO |
Spacer Arm | 10.9 Å |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...