DTME (dithio-bis-maleimidoethane), Pierce™
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DTME
22335
PIER22335EA
379
EUR
PIER22335
DTME (dithio-bis-maleimidoethane), Pierce™
DTME (dithio-bis-maleimidoethane)
Thermo Scientific Pierce DTME is a mid-length, maleimide crosslinker for reversible covalent conjugation between sulfhydryl groups (e.g., protein or peptide cysteines) by reduction of the disulfide bond in the center of the spacer arm.
- Reactive groups: maleimide (both ends)
- Reactive towards: sulfhydryl groups
- Long (13.3A), cleavable, sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 11-atom disulfide spacer arm
- Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
- Cleavable by reduction of disulfide spacer arm with DTT, TCEP or other reducing agent
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
C₁₂H₁₂N₂O₄S₂ Molecular Weight: 312,36 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01863385 CAS-Nummer: 71865-37-7 |
Spezifikation Testergebnisse
Product Line | Pierce™ |
Labeling Method | Chemical Labeling |
Crosslinker Type | Homobifunctional |
Reactive Moiety | Maleimide |
Spacer | Cleavable, Long |
Form | Solid |
Quantity | 50 mg |
Type | Crosslinker |
Solubility | DMF, DMSO |
Spacer Arm | 13.3 Å |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...