SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: SMPB

22416
PIER22416EA 249 EUR
PIER22416
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate), Pierce™
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)

Thermo Scientific Pierce SMPB is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length aromatic spacer arm (11.6 angstroms).


  • Reactive groups: NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Extended chain analog of MBS
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO)
  • Conjugates formed with SMPB are more stable in serum than SPDP conjugates


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₈H₁₆N₂O₆
Molecular Weight: 356,33 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00054961
CAS-Nummer: 79886-55-8

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Mid-Length
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR