L-Photo-Leucine, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Photo-Leucine, L-2-Amino-4,4'-azi-pentanoic acid

22610
PIER22610EA 546 EUR
PIER22610
L-Photo-Leucine, Pierce™
L-Photo-Leucine

Thermo Scientific Pierce L-Photo-Leucine is an analog of L-leucine that can be incorporated into proteins during synthesis and then light-activated to covalently crosslink interacting proteins in cells.


  • In vivo labeling—incorporate photo-reactive group into proteins using normal cellular machinery of live cells
  • In vivo crosslinking—find interacting proteins in the native cellular environment
  • Increased specificity—crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains
  • Efficient recovery—90% protein recovery in cell lysates after crosslinking
  • Compatible—crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6
  • Easy to use—reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark


Thermo Scientific L-Photo-Leucine is an analog of L-leucine that can be endogenously incorporated into the primary sequence of proteins during synthesis and then ultraviolet light (UV)-activated to covalently crosslink proteins within protein-protein interaction domains in their native environment within live cells. The powerful method enables both stable and transient protein interactions in cells to be studied and characterized without the use of completely foreign chemical crosslinkers and associated reagent solvents during the interaction experiment that can adversely aspects of the cell biology being studied.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Bestellen

Spezifikation Testergebnisse

Product Line Pierce™
Labeling Method Chemical Labeling, Metabolic Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Diazirine
Spacer Short
Form Solid
Quantity 100 mg
Type Crosslinker
Solubility Water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR