N-Ethylmaleimid (NEM), Pierce™

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-Ethylmaleinsäureimid

23030
PIER23030EA 193 EUR
PIER23030
N-Ethylmaleimid (NEM), Pierce™
N-Ethylmaleimid (NEM)

Thermo Scientific Pierce N-Ethylmaleimide (NEM) is a small compound that forms stable, covalent thioether bonds with sulfhydryls (e.g., reduced cysteines), enabling them to be permanently blocked to prevent disulfide bond formation.


  • Related applications: Protein Labeling & Crosslinking

Formel: C₆H₇NO₂
Molecular Weight: 125,13 g/mol
Siedepunkt: 210 °C (1013 hPa)
Schmelzpunkt: 46 °C
MDL: MFCD00005509
CAS-Nummer: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 13,03855

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR