Controlled Protein-Protein Crosslinking Kit, Pierce™
Lieferant: Thermo Fisher Scientific
The Pierce™ Controlled Protein-Protein Crosslinking Kit contains essential crosslinkers, modification reagents and buffers for conjugating antibodies, enzymes and other purified proteins needed as probes.
- Reliable non cleavable heterobifunctional crosslinking agent offers proven chemistry, highly stable intermediates and efficient formation of the target conjugate
- Includes disulfide (S-S) reductants and thiolation reagents
- Ellman’s Reagent provides the option to monitor reaction sufficiently
Lieferumfang: The kit includes Sulfo-SMCC crosslinking agent (2 mg); 10X activation/conjugation buffer (20 ml); BupH phosphate buffered saline (2 packs); 2-Mercaptoethylamine-HCI (6 mg); immobilised reductant (0,2 ml); SATA (2 mg); Dimethylformamide (DMF, 1 ml); Hydroxylamine-HCI (5 mg); dextran desalting columns (2×10 ml); column extender (2 each); cysteine-HCI (20 mg); and Ellman’s reagent (2 mg).