Salzsäure solution_SC 6N, Sequencing grade, Pierce™

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Hydrochloric acid solution, Muriatic acid

24308
PIER24308EA 82 EUR
PIER24308
Salzsäure solution_SC 6N, Sequencing grade, Pierce™
Salzsäure

Pierce™ Hydrochloric acid solution, sequencing grade, is purified and packaged to ensure a ninhydrin-negative blank on hydrolysis. This virtually eliminates exposure to laboratory atmospheres, fingerprints and other contaminants resulting from pipetting from bulk bottles.


  • Convenient, pre-scored ampoule packaging maintains reagent integrity
  • Hydrolyses peptides in 6 hours at 150 °C
  • Purified to give a ninhydrin-negative blank on hydrolysis

Formel: HCl
Molecular Weight: 36.46 g/mol
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
UN: 1789
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR