Pierce™ Glycoprotein Staining Kit

Lieferant: Thermo Fisher Scientific

24562
PIER24562EA 404 EUR
PIER24562
Pierce™ Glycoprotein Staining Kit
Färbemittel Elektrophorese-Färbemittel

The Pierce Glycoprotein Staining Kit is a convenient, fast, and sensitive colorimetric kit for staining glycosylated proteins in polyacrylamide gels or nitrocellulose membranes using the periodic acid-Schiff (PAS) method.


  • Specific for glycoproteins - PAS method specifically detects glycosylated proteins with sialic acid and other oxidisable carbohydrate groups
  • Protocol - the traditional three-reagent method has been streamlined to achieve optimal staining in less than two hours
  • Colorimetric - glycoproteins are detected as magenta bands against a faint pink or colorless gel background
  • Qualitative staining - detection limit depends on the extent of glycosylation and protein size and abundance; detects 0,625 ng avidin and 0.16 µg horseradish peroxidase per band
  • Controls included - complete kit includes positive and negative control proteins (HRP and STI, respectively)


The kit provides three essential reagents and a complete protocol to specifically stain glycosylated proteins (glycoproteins) that have been separated by polyacrylamide gel electrophoresis (PAGE). A gel or membrane containing separated proteins is treated with a periodate solution, which oxidises cis-diol sugar groups in glycoproteins. The resulting aldehyde groups are detected through the formation of Schiff-base bonds with a reagent that produces magenta bands. Traditionally called the periodic acid-Schiff (PAS) reagent method, this staining technique is made easier by the convenient packaging of essential reagents in one kit with easy-to-follow instructions.


Contents
Sufficient for: 10 mini gels (SDS-PAGE)
• Oxidation reagent, 2,5 g powder to make 250 ml
• Reduction reagent, 1,25 g powder to make 250 ml
• Positive control (Horseradish peroxidase), 1 mg
• Negative control (Soybean trypsin inhibitor), 1 mg
• Glycoprotein stain, 250 ml


Detection location: In-blot detection, in-gel detection
Detection method: Colorimetric
Label or dye: Fuchsin sulfite
Target molecule: Proteins (Glycoproteins)


Bestellinformation: The 6xHis protein tag staining kit (PI24575) and phosphoprotein staining kit (PI24550) come with the appropriate reagent and control sets, which can be purchased separately.


Bitte beachten Store all components at 4 °C. For research use only. Not for use in diagnostic procedures.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR