Pierce™ 6xHis Protein Tag Stain Reagent Set

Lieferant: Thermo Fisher Scientific

24570
PIER24570EA 456 EUR
PIER24570
Pierce™ 6xHis Protein Tag Stain Reagent Set
Färbemittel Elektrophorese-Färbemittel

The Thermo Scientific Pierce 6xhis protein tag stain reagent set is a fluorescent stain for preferential detection of histidine-tagged (his-tagged) recombinant fusion proteins in protein polyacrylamide gels.


  • Two to three times faster than western blotting get results sooner and save hours of valuable research time
  • Detects directly on the gel can eliminate the need for a membrane transfer and western blot step for the detection of at least 0,2 µg of 6xhis-tagged protein
  • Ready-to-use, two-reagent formula no mixing, no diluting and no fuss; guarantees a simple-to-perform, mistake-free detection protocol
  • Specific fluorescent detection of 6xHis-tagged proteins see only what you want to see (CCD camera recommended for low abundance proteins; UV-transilluminator for abundant proteins)
  • Compatible with double-staining follow his-tagged protein staining with Thermo Scientific GelCode blue stain reagent for a total protein profile determination


The unique fluorescent stain specifically detects histidine-tagged fusion proteins that have been electrophoresed in polyacrylamide gels (e.g., SDS-PAGE). With this stain, expression of His-tagged recombinant proteins can be assessed directly in-gel, eliminating membrane transfer and Western blotting steps that are typically used to confirm purity and yield of tagged proteins. Although the lower limit of detection depends on which UV lamp (300 nm) and detection equipment (CCD camera best) are used, the stain is convenient for in-process assessment of his-tagged protein production and is compatible with subsequent staining with coomassie dye and other total protein stains.


Includes: Stain and developer solutions (500 ml each)
Requires: UV lamp source (300 nm) and CCD camera


Detection location: In-gel detection
Detection method: Fluorescent
Label or dye: Proprietary mix
Target molecule: Proteins (His-tagged)


Store components at room temperature. Product is shipped at ambient temperature.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR