Protein gel stain, GelCode™ SilverSNAP® silver stain II, Pierce™
Lieferant: Thermo Fisher Scientific
GelCode™ SilverSNAP® Silver Stain II is a fast, flexible stain that delivers consistently reliable staining performance.
- Low and uniform background
- Detection as low as 0,25 ng of protein
- Staining can be completed in under 50 minutes
- Compatible with a wide assortment of homemade and precast gels
- Flexible protocol without altering sensitivity or background
Suitable for use with 1- or 2-D PAGE and IEF gels and used to stain DNA or RNA following electrophoresis.