SATP (N-Succinimidyl 3-(Acetylthio)propionate), Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: SATP

26100
PIER26100EA 125 EUR
PIER26100
SATP (N-Succinimidyl 3-(Acetylthio)propionate), Pierce™
SATP (N-Succinimidyl 3-(Acetylthio)propionate)

SATP (and SATA) are reagents for introducing protected sulfhydryls into proteins, peptides and other molecules. for covalent modification of primary amines and addition of a protected sulphhydryl group.


  • Adds a protected sulfhydryl that can be deprotected by hydroxylamine
  • Allows long-term storage of the sulfhydryl-modified molecule
  • Offers more steric freedom than SATA for the unmasked sulfhydryl group
  • Forms cleavable disulfide bonds with other sulfhydryl-containing molecules
  • Reacts with primary amines (eg. lysine residues proteins) to form stable amide bonds
  • Preserves protein activity with its mild, non-denaturing reaction cdonditions


SATP (and SATA) are the N-hydroxysuccinimide (NHS) esters of S-acetylthioacetic and propionic acid. A stable, covalent amide bond is formed from the reaction of the NHS ester with primary amines. The amine reacts with the NHS ester by nucleophilic attack, with N-hydroxysuccinimide being released as a by-product. Deprotection (deacylation) to generate a sulfhydryl for use in cross-linking and other applications is accomplished using hydroxylamine·HCl.

Formel: C₉H₁₁NO₅S
Molecular Weight: 245.25 g/mol
MDL: MFCD00133023
CAS-Nummer: 84271-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR