Hydroxylammoniumchlorid, Pierce™
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Hydroxylamin-Hydrochlorid, Chlorhydrathydroxylamin, Hydroxylamin Hydrochlorid, Hydroxylamin HCl
26103
PIER26103EA
70.25
EUR
PIER26103
Hydroxylammoniumchlorid, Pierce™
Hydroxylammoniumchlorid
Thermo Scientific Pierce Hydroxylamine-HCl has many uses, which include cleaving Asn-Gly peptide bonds and deprotecting (exposing) sulfhydryls on SATA-modified molecules for conjugation.
- Related applications: Protein Labeling & Crosslinking
Formel:
NH₂OH·HCl Siedepunkt: 306 °C (1013 hPa) Schmelzpunkt: 151 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III Merck Index: 13,04853 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...