3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)

Lieferant: Thermo Fisher Scientific

Synonyme: CHAPS

28300
PIER28300EA 217 EUR
PIER28300
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat (CHAPS)
Formel: C₃₂H₅₈N₂O₇S
Molecular Weight: 614,89 g/mol
Schmelzpunkt: 116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00081083
CAS-Nummer: 75621-03-3
Merck Index: 13,02052

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR