N-Octyl-β-D-glucopyranosid nichtionisches Detergens, Pierce™

Lieferant: Thermo Fisher Scientific
Achtung

28310
PIER28310EA 542 EUR
PIER28310
N-Octyl-β-D-glucopyranosid nichtionisches Detergens, Pierce™
N-Octyl-β-D-glucopyranosid

Thermo Scientific Octyl-beta-Glucoside is a nonionic detergent that has been widely used for membrane protein solubilization.


  • A low-molecular weight, nonionic detergent
  • Effective for membrane protein solubilization
  • Less stable and less commonly used than octylthioglucoside (OTG)
  • Can be removed from solution by dialysis
  • Optically clear; low absorbance at 280 nm


White, free-flowing powder, free of foreign material. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.

Formel: C₁₄H₂₈O₆
Schmelzpunkt: 110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063288
CAS-Nummer: 29836-26-8
EINECS: 249-887-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR