Natriumdodecylsulfat (SDS) C12, Pierce™
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Natriumlaurylsulfat, Schwefelsäuredodecylester Natriumsalz, Laurylsulfat Natriumsalt, Dodecylnatriumsulfat, Dodecylschwefelsäure Natriumsalz
Thermo Scientific Sodium Dodecyl Sulfate (C12) is a highly purified form of SDS that is especially suited for protein experiments requiring tightly controlled solubility parameters.
- Popular anionic detergent for protein electrophoresis and solubilization
- Component of many cell lysis buffer formulations
- Specially purified formulation is>98% C12 alkyl sulfate
- Contains low levels of hexadecyl sulfate (C16), which inhibits protein renaturation
Sodium dodecyl sulfate (SDS) is an alkyl sulfate, anionic detergent used frequently in protein electrophoresis and protein solubilization methods. It is a component of many cell lysis buffer formulations, including RIPA buffer. Typical SDS or lauryl sulfate preparations are mixtures of several different chain lengths (C10 to C18). However, protein renaturation and other specialized methods require very specific surfactant and buffer conditions. This particular formulation of SDS is highly purified to eliminate nearly all secondary chain lengths besides C12. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25 ˚C.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
CH₃(CH₂)₁₁OSO₃Na Molecular Weight: 288.38 g/mol Schmelzpunkt: 204…207 °C Dichte: 1.1 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036175 CAS-Nummer: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1 II Merck Index: 14,08636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...