PBS-Puffer-Lösung 20fach konzentriert (phosphatgepufferte Salzösung)
Lieferant: Thermo Fisher Scientific
28348
PIER28348EA 151 EUR
PIER28348
PBS-Puffer-Lösung 20fach konzentriert (phosphatgepufferte Salzösung)
Puffer Elektrophorese-Puffer Waschpuffer
Our variety of Pierce Concentrated Buffer stock solutions are ready to use without having to weigh and dissolve dry ingredients or adjust the pH with concentrated acid or base. Simply dilute the liquid stock solution with pure water and proceed with your experiment.
  • Convenient—no weighing, no pH adjustment, no need to stock individual components
  • Improved accuracy—ready-to-use formula, easily diluted without weighing chemical components
  • Consistent quality—our quality control ensures that every prep will yield the same, consistent buffer
20× PBS Tween-20 can be used to prepare PBS-Tween (PBS-T) wash buffers for ELISA, western, and other immunoassays, as well as blocking buffer for plate-based assays. When diluted to 1× with water, 20× PBS Tween-20 makes 10 mm sodium phosphate, 0,15 M NaCl, 0,05% Tween™ 20 buffer at pH 7,5.