Octyl-β-D-thioglucopyranosid nichtionisches Detergens, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

28351
PIER28351EA 341 EUR
PIER28351
Octyl-β-D-thioglucopyranosid nichtionisches Detergens, Pierce™
Octyl-β-D-thioglucopyranosid

Octylthioglucoside (OTG) is a low-molecular weight non ionic detergent that is used for cell lysis and non denaturing protein solubilisation.


  • Resistant to beta-D-glucoside glucohydrolase degradation
  • Unaffected by beta-glucosidase enzyme
  • More stable than octyl beta-glucoside
  • Can be removed from solution by dialysis
  • Optically clear, low absorbance at 280 nm

Formel: C₁₄H₂₈O₅S
Molecular Weight: 308,44 g/mol
MDL: MFCD00012189
CAS-Nummer: 85618-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR