TAE buffer (20X), Pierce™
Lieferant: Thermo Fisher Scientific
28354
PIER28354EA 116 EUR
PIER28354
TAE buffer (20X), Pierce™
Puffer Elektrophorese-Puffer Laufpuffer
20X TAE buffer is a space-saving stock solution that is ideal for casting and preparing Tris-Acetate EDTA running buffer used for agarose gel electrophoresis of nucleic acids.
When diluted to 1X with water, yields 00,4 M Tris, 0,04 M acetate, 1 mM EDTA, pH 8,2 to 8,4.