Wash buffer, Pierce™
Lieferant: Thermo Fisher Scientific
28341 28344 28346 28352 28360
PIER28341EA 121 EUR
PIER28341 PIER28344 PIER28346 PIER28352 PIER28360
Wash buffer, Pierce™
Puffer Elektrophorese-Puffer Waschpuffer
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
  • Ready to use without having to reconstitute with ultrapure water
  • Eliminates the possibility of powder remaining in a packet
  • 10X or 20X concentration eliminates time spent waiting for powder to dissolve
  • Storage as concentrated stock minimizes bench space needed for large volume solutions