D(+)-Biotin ≥97,5%, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: (3aS,4S,6aR)-Biotin, Vitamin H, Vitamin B8, 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H- thieno[3,4-d]imidazol-4-yl]pentansäure, Vitamin B7, Coenzym R

29129
PIER29129EA 92.5 EUR
PIER29129
D(+)-Biotin ≥97,5%, Pierce™
D(+)-Biotin

Biotin, also known as vitamin H, is a small molecule (MW 244.3) that is present in tiny amounts in all living cells and is critical for a number of biological processes.


The valeric acid side chain of the biotin molecule can be derivatized in order to incorporate various reactive groups that are used to attach biotin to other molecules. In the context of immunohistochemistry (IHC), biotin is conjugated to antibodies or to the enzyme reporters used to detect target antigens.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₀H₁₆N₂O₃S
Molecular Weight: 244,31 g/mol
Schmelzpunkt: 232…233 °C
Dichte: 1,41 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00005541
CAS-Nummer: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR