Pierce™ Recombinant Protein G, Biotinylated
Lieferant: Thermo Fisher Scientific
29988
PIER29988EA
274
EUR
PIER29988
Pierce™ Recombinant Protein G, Biotinylated
Protein-Aufbereitungs
Proteinreinigungs-Reagenzien
Biotinylated recombinant Protein G can be used to probe and detect mouse and human antibodies, especially IgG isotypes, in western blotting, ELISA, IHC and other immunoassay protocols.
- Contains two Fc-binding domains per protein
- Does not bind human IgM, IgD or IgA
- Efficient than Protein A for mouse (IgG1), human (IgG3) and rat, goat and cow antibodies
- Less efficient than Protein A for guinea pig, pig, dog and cat