Recombinant protein L, biotinylated, Pierce™
Lieferant: Thermo Fisher Scientific
29997
PIER29997EA
307
EUR
PIER29997
Recombinant protein L, biotinylated, Pierce™
Protein-Aufbereitungs
Proteinreinigungs-Reagenzien
Pierce™ Biotinylated Recombinant Protein L can be used to probe and detect mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.
- Contains four immunoglobulin binding domains per protein
- Binds members of all classes of Ig that contain kappa light chains
- Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen binding site.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...