Recombinant protein A/G, alkaline phosphatase conjugated, Pierce™

Lieferant: Thermo Fisher Scientific

32391
PIER32391EA 314 EUR
PIER32391
Recombinant protein A/G, alkaline phosphatase conjugated, Pierce™
Protein-Aufbereitungs Proteinreinigungs-Reagenzien

Pierce™ Recombinant Alkaline Phosphatase Conjugated Protein A/G is useful to probe and detect antibodies, especially IgG isotypes, of many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.


  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognised by either Protein A or Protein G


Gene fusion of the Fc-binding domains of Protein A and Protein G has resulted in production of a structurally and functionally chimeric protein with broader binding than either Protein A or Protein G alone. During fusion, the Protein G gene sequence coding for the serum albumin-binding site is eliminated. This protein is expressed in E. coli and secreted into the surrounding medium during fermentation. The product obtained is consistent in quality and yield because the bacterial host is engineered to be deficient in major proteolytic activities.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR