Recombinant protein A, peroxidase conjugated, Pierce™

Lieferant: Thermo Fisher Scientific

32400
PIER32400EA 219 EUR
PIER32400
Recombinant protein A, peroxidase conjugated, Pierce™
Protein-Aufbereitungs Proteinreinigungs-Reagenzien

Pierce™ Recombinant Peroxidase Conjugated Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.


  • Contains four Fc-binding domains per protein
  • More efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies


Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. Recombinant Protein A is produced in E. coli and functions essentially the same as native Protein A. The Protein A molecule contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8,2, although binding is also effective at neutral or physiological conditions (pH 7,0 to 7,6).

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR