Blocker™ Blocking Buffer, BLOTTO in TBS
Lieferant: Thermo Fisher Scientific
37530
PIER37530EA 178 EUR
PIER37530
Blocker™ Blocking Buffer, BLOTTO in TBS
Puffer Elektrophorese-Puffer Blockier-Puffer
Blocker™ BLOTTO is a ready-to-use 5% solution of non fat powdered milk in Tris-buffered saline for Western blot, ELISA, IHC and other detection methods.
  • Convenient as can be diluted as needed
  • Easy to use, formulated with anti-foaming agent and thimerosal-free preservative
  • Flexible may be used for multiple applications
Bitte beachten For research use only. Not for use in diagnostic procedures.