StartingBlock™ Blocking Buffer
Lieferant: Thermo Fisher Scientific
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
- Compatible with many detection systems (Western blot, ELISA and IHC)
- Short blocking times
- High signal:noise ratio
- Strip and reprobe without reblocking
- Available ready to use 1X formulations in PBS and TBS
- For research use only
- Not for use in diagnostic procedures
It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.