Pierce™ Blocking Buffers, Protein Free
Lieferant: Thermo Fisher Scientific
37570
PIER37570EA 251 EUR
PIER37570
Pierce™ Blocking Buffers, Protein Free
Puffer Elektrophorese-Puffer Blockier-Puffer
These protein-free blocking buffers are either a PBS or a TBS formulation of a non protein compound that provides effective blocking for membrane based and plate based protein detection methods, resulting in extremely low background.
  • Streptavidin friendly that is absolutely free of biotin
  • Effective for all kinds of protein detection systems
  • Optimized and validated in many protein methods to provide high signal-to-noise ratio
  • Protein free blocker that minimizes or eliminates cross reactivity associated with protein-based blocking buffers
These buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, western blotting, glass slide arrays and other applications.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at room temperature. After opening, store product at 4˚C.