HRP substrate, 1-Step™ ABTS Substrate Solution
Lieferant: Thermo Fisher Scientific
37615
PIER37615EA
153
EUR
PIER37615
HRP substrate, 1-Step™ ABTS Substrate Solution
Tests
ELISAs
The 1-Step™ ABTS (2,2'-azinobis [3-ethylbenzthiazoline-6-sulfonic acid] diammonium salt) is a ready-to-use, water-soluble ELISA substrate for horseradish peroxidase. This substrate yields a green product with absorbance maxima at 410 nm and 650 nm. Adding SDS to the substrate stops the reaction without changing the colour.