L(+)-Cystein Hydrochlorid, Pierce™
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid
44889
PIER44889EA
92.75
EUR
PIER44889
L(+)-Cystein Hydrochlorid, Pierce™
L(+)-Cystein Hydrochlorid
Thermo Scientific Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent or as an additive in protein refolding experiments.
- Related applications: Protein Labelling and Crosslinking
For Research Use Only. Not for use in diagnostic procedures.
Verpackung: For research use only. Not for use in diagnostic procedures.
Formel:
HSCH₂CH(NH₂)COOH·HCl Schmelzpunkt: 175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064553 CAS-Nummer: 52-89-1 EINECS: 200-157-7 Merck Index: 13,02810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...