Pierce™, CarboxyLink™ Immobilisation Kit
Lieferant: Thermo Fisher Scientific
44899
PIER44899EA
708
EUR
PIER44899
Pierce™, CarboxyLink™ Immobilisation Kit
Protein-Aufbereitungs
Protein Purification Kits
CarboxyLink™ immobilisation kit is for covalent immobilisation of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.
- Ideal for unmodified peptides, immobilises peptides with high capacity and various orientations without steric hindrance
- DADPA-activated crosslinked 4% beaded agarose acting as a CarboxyLink™ coupling resin
- With stable linkage, flexible and gentle coupling conditions
Lieferumfang: Kit includes CarboxyLink™ columns, 2 ml (5 columns), EDC crosslinker (5×60 mg), MES buffered saline pack (makes 500 ml), wash solution (120 ml), and column accessories.